Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:14174-09-5
:
Formula:C₂₄H₃₂O₈
MDL Number:MFCD00005101
:448.51 g/mol
:M-Clarity™ quality level = MQ200
EINECS:238-027-7
Specification Test Results
- Melting Point:100 - 103 °C
- Molar Mass:448.52 g/mol