Order Entry
Germany
ContactUsLinkComponent
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) for synthesis, Sigma-Aldrich®
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.007 g/l (25 °C)
  • Melting Point:
    158 - 161 °C
  • Molar Mass:
    360.41 g/mol

Specification Test Results

Related Information