
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) 98+%
:14187-32-7
:
:C₂₀H₂₄O₆
:MFCD00005098
:360.41 g/mol
:380…384 °C (679 mmHg)
:160…164 °C
:Ambient
:238-041-3
:GHS07
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Melting point:160 to 164 °C
- HPLC:≥98.0 %
- NMR (1H-NMR):Consistent with structure