9-Aminoacridine hydrochloride monohydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:52417-22-8
:
Formula:C₁₃H₁₁ClN₂·1H₂O
MDL Number:MFCD00150071
:248.71 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:205-145-5
UN:2811