Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, 2-Isobutyl-3-methoxypyrazine solution
Supelco® Organic Reference Standard, 2-Isobutyl-3-methoxypyrazine solution
Catalog # 47528-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, 2-Isobutyl-3-methoxypyrazine solution
Catalog # 47528-U
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Certified reference material
  • CAS No.:
    24683-00-9
  • Description:
    2-Isobutyl-3-methoxypyrazine solution
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    11 °C

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311