Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acridine-[D9]
Acridine-[D₉] (98% D)
Acridine-[D₉] (98% D)
Acridine-[D₉] (98% D)
Supplier:
LGC Standards PROMOCHEM
Certificates
Expand image
Acridine-[D₉] (98% D)
Supplier:
LGC Standards PROMOCHEM
About this item
CAS Number:
34749-75-2
:
Formula:
C₁₃D₉N
:
188.15 g/mol
UN:
2713
ADR:
6.1,III