Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Trimethylsilyl)propionic acid
3-(Trimethylsilyl)propionic acid
3-(Trimethylsilyl)propionic acid
3-(Trimethylsilyl)propionic acid
Supplier:
CHEMPUR SILANE
Certificates
Expand image
3-(Trimethylsilyl)propionic acid
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
5683-30-7
:
Formula:
C₆H₁₄O₂Si
MDL Number:
MFCD00002761
:
146.26 g/mol
EINECS:
227-145-4