


Silver nitrate 0.01 mol/l (0.01 N) in methanol
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:1992
ADR:3,II
:GHS02
:GHS06
:GHS08
Frequently Bought Together