Order Entry
Germany
ContactUsLinkComponent
Silver nitrate 0.001 mol/l (0.001 N) in aqueous solution
Silver nitrate 0.001 mol/l (0.001 N) in aqueous solution
Catalog # KRAF16092.3700
Supplier:  Bernd Kraft
CAS Number:  
undefined
Silver nitrate 0.001 mol/l (0.001 N) in aqueous solution
Catalog # KRAF16092.3700
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414