Silver nitrate 0.001 mol/l (0.001 N) in aqueous solution
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
Frequently Bought Together
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Aquaresist Water Bath Protective Agent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection