Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethoxy(diisopropyl)silane
Dimethoxy(diisopropyl)silane
Dimethoxy(diisopropyl)silane
Dimethoxy(diisopropyl)silane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Dimethoxy(diisopropyl)silane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
18230-61-0
:
Formula:
C₈H₂₀O₂Si
MDL Number:
MFCD00239343
:
176.33 g/mol
EINECS:
421-540-7