Order Entry
Germany
ContactUsLinkComponent
Ammonium nickel(II) sulphate hexahydrate, Sigma-Aldrich®
Ammonium nickel(II) sulphate hexahydrate, Sigma-Aldrich®
Catalog # SIAL574988-25G
Supplier:  Merck
CAS Number:  
Ammonium nickel(II) sulphate hexahydrate, Sigma-Aldrich®
Catalog # SIAL574988-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :394.99 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:239-793-5
CAS Number:7785-20-8
Formula:H₈N₂NiO₈S₂·6H₂O
MDL Number:MFCD00150260