Order Entry
Germany
ContactUsLinkComponent
 
Aluminium tri-sec-butanolate for synthesis, Sigma-Aldrich®
 :  Merck
 
Aluminium tri-sec-butanolate for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    246.32 g/mol
  • Boiling Point:
    180 - 182 °C (5 hPa)
  • Vapor Pressure:
    23 hPa (195 °C)
  • Flash Point:
    26 °C
  • Explosion Limit:
    1.7 - 9.8 %(V)
  • Density:
    0.96 g/cm3 (20 °C)
  • Ignition Point:
    406 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid