Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium tri-sec-butanolate
Aluminium tri-sec-butanolate ≥95%
Aluminium tri-sec-butanolate ≥95%
:
Thermo Fisher Scientific
Aluminium tri-sec-butanolate ≥95%
:
Thermo Fisher Scientific
:
2269-22-9
:
:
C₁₂H₂₇AlO₃
:
MFCD00009327
:
246.33 g/mol
:
200…206 °C (30 mmHg)
:
0.967
:
27 °C (82 °F)
:
Ambient
:
218-871-2
:
1992
:
3,III
:
GHS02
:
GHS07
msds_ALFA030721
Thermo Scientific Acroseal 25 Years