Aluminium tri-sec-butanolate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2269-22-9
:
Formula:C₁₂H₂₇AlO₃
MDL Number:MFCD00009327
:246.33 g/mol
Boiling Pt:200…206 °C (30 mmHg)
Density:0.96 g/cm³
Flash Pt:27 °C
Storage Temperature:Ambient
EINECS:218-871-2
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear to slightly hazy colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4380 to 1.4400 (20°C, 589 nm)
- Assay:≥96.0 % (ex Al)