Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Cyclohexane, Technical Grade

Flammable
Health
Irritant
Environment
Supplier:   Bernd Kraft
Cyclohexane, Technical Grade
Supplier:  Bernd Kraft
Certificates
Cyclohexane, Technical Grade
Cyclohexane, Technical Grade

4 Options Available Below

Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 42,90 €
    42,90 €
    1 * 2.500 mL
       42,90 €

  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 69,70 €
    69,70 €
    1 * 5 L
       69,70 €

  • Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 113,00 €
    113,00 €
    1 * 10 L
       113,00 €

  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 250,00 €
    250,00 €
    1 * 25 L
       250,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.