


Methanol for LC-MS
KRAF22490.4700
:
- Pack type:Glass bottle
- Pk:2.500 mL
- Seq:2895
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
:
:32.04 g/mol
:13,05984
:64.6 °C (1013 hPa)
:–98 °C
:0.7918 g/cm³ (20 °C)
:11 °C (closed cup)
:Ambient
:200-659-6
:1230
:3,II
:01-2119433307-44
:GHS02
:GHS06
:GHS08
:67-56-1
:CH₃OH
:MFCD00004595