


Methanol for residue analysis
Catalog # KRAF17655.4700
Lieferant: Bernd Kraft
CAS-Nummer:
Spezifikationen
- Pack type:Glass bottle
- Pk:2.500 mL
- Seq:2895
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Spezifikationen
Über diesen Artikel
:
:32.04 g/mol
Merck-Index:13,05984
Siedepunkt:64.6 °C (1013 hPa)
Schmelzpunkt:–98 °C
Dichte:0.7918 g/cm³ (20 °C)
Flammpunkt:11 °C (closed cup)
Lagertemperatur:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
CAS-Nummer:67-56-1
Formel:CH₃OH
MDL-Nummer:MFCD00004595