0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Sulphuric acid
Sulphuric acid 0.05 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
Sulphuric acid 0.05 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
:
Bernd Kraft
Sulphuric acid 0.05 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
:
Bernd Kraft
:
7664-93-9
:
:
H₂SO₄
:
MFCD00064589
:
98.08 g/mol
:
1.01 g/cm³ (20 °C)
:
Ambient
:
231-639-5
:
2796
:
8,II