Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acetic acid
Acetic acid ≥90%, Technical Grade
Acetic acid ≥90%, Technical Grade
:
Bernd Kraft
Acetic acid ≥90%, Technical Grade
:
Bernd Kraft
:
64-19-7
:
:
H₃CCOOH
:
MFCD00036152
:
60.05 g/mol
:
Ambient
:
200-580-7
:
2789
:
8,II
:
GHS02
:
GHS05