
Ammonia solution 2 mol/l (2 N)
:1336-21-6
:7664-41-7
:NH₃
:MFCD00066650
:35.05 g/mol
:Ambient
:215-647-6
:2672
:8,III
:GHS07
Frequently Bought Together
Ethylene glycol ≥99.0% Reag. Ph. Eur.
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals