

Silver nitrate 0.4 mol/l (0.4 N) in aqueous solution
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:3264
ADR:8,II
:GHS05
:GHS09
Frequently Bought Together