Order Entry
Germany
ContactUsLinkComponent
Biphenyl (PCB No. 0)
Irritant
Environment
Biphenyl (PCB No. 0)
Catalog # PROOCILULM17100.5
CAS Number:  
Irritant
Environment
Biphenyl (PCB No. 0)
Catalog # PROOCILULM17100.5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :154.21 g/mol
Merck Index:13,03346
Boiling Pt:255 °C (1013 hPa)
Melting Pt:68…70 °C
Density:1.175 g/cm³ (20 °C)
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:202-163-5
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:92-52-4
Formula:C₁₂H₁₀
MDL Number:MFCD00003054