Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Sodium hydroxide 4% in aqueous solution, Technical Grade

Corrosive
Supplier:   Bernd Kraft
Sodium hydroxide 4% in aqueous solution, Technical Grade
Supplier:  Bernd Kraft
Certificates
Sodium hydroxide 4% in aqueous solution, Technical Grade

3 Options Available Below

Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 61,00 €
    61,00 €
    1 * 5 L
       61,00 €

  • Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 98,80 €
    98,80 €
    1 * 10 L
       98,80 €

  • Plastic bottle
    20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L - 145,00 €
    145,00 €
    1 * 20 L
       145,00 €