Ammonium iron(II) sulphate 0.2 mol/l (0.2 N) in aqueous solution for COD determination
Catalog # KRAF01249.3000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:284.05 g/mol
Storage Temperature:Ambient
EINECS:233-151-8
UN:3264
ADR:8,II
CAS Number:10045-89-3
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039