Order Entry
Germany
ContactUsLinkComponent
Ammonium iron(II) sulphate 0.25 mol/l (0.25 N) in aqueous solution for COD determination
Ammonium iron(II) sulphate 0.25 mol/l (0.25 N) in aqueous solution for COD determination
Catalog # KRAF01049.3000
Supplier:  Bernd Kraft
CAS Number:  
Ammonium iron(II) sulphate 0.25 mol/l (0.25 N) in aqueous solution for COD determination
Catalog # KRAF01049.3000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :284.05 g/mol
Storage Temperature:Ambient
EINECS:233-151-8
UN:3264
ADR:8,II
CAS Number:10045-89-3
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039