Order Entry
Germany
ContactUsLinkComponent
Celite® 545, filter aid NF, acid-washed
Health
Celite® 545, filter aid NF, acid-washed
Catalog # VWRS6095.5000
Supplier:  VWR Collection
CAS Number:  
Health
Celite® 545, filter aid NF, acid-washed
Catalog # VWRS6095.5000
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :60.08 g/mol
EINECS:272-489-0
 :GHS08
CAS Number:68855-54-9
Formula:SiO₂
MDL Number:MFCD00132803