2-Iodothiophene
Catalog # MOLEM43009263
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:210.04 g/mol
Boiling Pt:182 °C (1013 hPa)
Melting Pt:–1 °C
Density:2.0645 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:222-342-1
CAS Number:3437-95-4
Formula:C₄H₃IS
MDL Number:MFCD00005424