




3-Chlorothiophene ≥98% may contain up to 2% DMF
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17249-80-8
:
Formula:C₄H₃ClS
MDL Number:MFCD00043887
:118.59 g/mol
Boiling Pt:137…139 °C
Density:1.276
Flash Pt:37 °C (98 °F)
UN:1992
ADR:3,III
:GHS06
:GHS08
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient