Order Entry
Germany
Orders LinkContactUsLinkComponent
2,3,5-Collidine

2,3,5-Collidine

Glass bottle for solids, 25 g
Catalog # MOLE15131983-25G
Irritant
Marketsource
Supplier:   Molekula
2,3,5-Collidine
Catalog # MOLE15131983-25G
Supplier:  Molekula
Marketsource
CAS Number:  695-98-7
2,3,5-Collidine

Glass bottle for solids, 25 g

CAS Number:  695-98-7
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2,3,5-Trimethylpyridine
  • CAS Number:695-98-7
  • Formula:C₈H₁₁N
  • MDL Number:MFCD00126840
  • Molecular Weight:121.18 g/mol
  • Boiling Pt:182 °C (1013 hPa)
  • Density:0.9508 g/cm³ (0 °C)
  • EINECS:211-786-1
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications