2-Vinylpyridine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-69-6
:
Formula:C₇H₇N
MDL Number:MFCD00006355
:105.14 g/mol
Boiling Pt:159 °C (1013 hPa)
Melting Pt:–50 °C
Density:0.9735 g/cm³ (20 °C)
Flash Pt:51 °C
Storage Temperature:Freezer
UN:3073
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:202-879-8