Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyridine analytical reagent

Pyridine analytical reagent

Flammable
Irritant
Supplier:   Bernd Kraft
Pyridine analytical reagent
Supplier:  Bernd Kraft
Certificates
Pyridine analytical reagent
Pyridine analytical reagent

3 Options Available Below

Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:13,08060
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.983 g/cm³ (20 °C)
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 106,00 €
    106,00 €
    1 * 500 mL
       106,00 €

  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 366,00 €
    366,00 €
    1 * 2.500 mL
       366,00 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 173,00 €
    173,00 €
    1 * 1 L
       173,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.