Stirring shafts, fan shaped, BOLA
Stirrer shafts made of PTFE-jacketed stainless steel and the fan shaped stirring unit completely made of PTFE.
- Suitable for stirring in narrow mouth vessels or in vessels with ground joint openings
- Chemical resistant
- Temperature resistance from –200 to +250 °C