Order Entry
Germany
ContactUsLinkComponent
2-Chloropyridine
2-Chloropyridine
Catalog # EHERC11503700
Supplier:  EHRENSTORFER
CAS Number:  
2-Chloropyridine
Catalog # EHERC11503700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :113.55 g/mol
Boiling Pt:166 °C (1013 hPa)
Melting Pt:–46 °C
Density:1.205 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-646-3
UN:2822
ADR:6.1,II
CAS Number:109-09-1
Formula:C₅H₄ClN
MDL Number:MFCD00006228