5-Bromo-2-iodopyridine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:223463-13-6
:
Formula:C₅H₃BrIN
MDL Number:MFCD00691506
:283.89 g/mol
Melting Pt:112…113 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:111 to 115 °C
- GC:≥97.5 %