Order Entry
Germany
Orders LinkContactUsLinkComponent
Tetrahydrofuran | Avantor

Tetrahydrofuran

Catalog #
Entzündlich
Gesundheit
Reizend
Tetrahydrofuran
Catalog #
Supplier:  
CAS Number:  109-99-9
Tetrahydrofuran
Tetrahydrofuran
CAS Number:  109-99-9
Product Details & Documents
Synonyme:  Butylene oxide, THF, Diethylene oxide
  • CAS-Nummer:109-99-9
  • Formel::C₄H₈O
  • MDL-Nummer:MFCD00005356
  • Molekulargewicht:72.11 g/mol
  • Merck-Index:12,09351
  • Siedepunkt:66 °C (1013 hPa)
  • Schmelzpunkt:–108.5 °C
  • Dichte:0.888 g/cm³ (20 °C)
  • Flammpunkt:–21.5 °C
  • Lagertemperatur:Ambient
  • EINECS:203-726-8
  • UN:2056
  • ADR:3,II
  • REACH:05-2114615188-48