Tetrahydrofuran
EHERC17405700
:
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
:
:72.11 g/mol
:12,09351
:66 °C (1013 hPa)
:–108.5 °C
:0.888 g/cm³ (20 °C)
:–21.5 °C
:Ambient
:203-726-8
:2056
:3,II
:05-2114615188-48
:GHS02
:GHS08
:GHS07
:109-99-9
:C₄H₈O
:MFCD00005356