About this item
Rotavapor® R-210 rotary evaporator for rapid and complex distillation of all types of solvents. Works with evaporator flasks of 50 to 4000 ml. The R-210 is equipped with an electric lifting and lowering device and automatic lifting of the evaporator flask in the event of a power failure.
- Electronically adjustable speed
- Heating bath designed to heat water or oil within a temperature range of 20 to 180 °C
- Heating bath comes with a digital temperature display fitted as standard
Rotavapor® R-210 Advanced
The R-210 Advanced comes with a V-850 vacuum controller that is used to control a V700 or V710 pump and which has a timer function and program memory. Rotating and lifting can be controlled by the controller. The integrated library containing 43 solvents, which can be updated via USB, facilitates fast and efficient evaporation.
Rotavapor® R-210 Professional**
The R-210 Professional includes the V-855 vacuum controller. You can choose different settings, such as continuous vacuum, pressure gradients, timer functions or complete automatic distillation. Integrated EasyVac and LabVac functions. Rotating and lifting can be controlled. The integrated library containing 43 solvents, which can be updated via USB, facilitates fast and efficient evaporation. Please contact your local Avantor sales office for details.
Glassware
Glassware A, diagonal condenser: For standard distillations, can be used in a confined space, evaporator flask feed.
Glassware V, vertical condenser: For standard distillations, evaporator flask feed, connection for vapour temperature sensor, automatic distillation possible with the V-855 vacuum controller.
Glassware C, dry ice condenser: For distilling solvents with low boiling points, evaporator flask feed, no cooling liquid necessary.
Glassware S, vertical condenser with shut-off valve: For standard distillations, evaporator flask feed, connection for vapour temperature sensor, automatic distillation possible with V-855 vacuum controller.
Glassware E, descending condenser with expansion vessel: Ideal for distilling substances that have a tendency to foam and that take a long time to reach boiling point, evaporator flask feed, connection for vapour temperature sensor.
Glassware BY, intensive condenser: Vertical condenser with double mantle, additional joint on the top of the condenser to allow expansion, evaporator flask feed, connection for vapour temperature sensor.
The R-210 Professional includes the V-855 vacuum controller. You can choose different settings, such as continuous vacuum, pressure gradients, timer functions or complete automatic distillation. Integrated EasyVac and LabVac functions. Rotating and lifting can be controlled by the controller. The integrated library containing 43 solvents, which can be updated via USB, facilitates fast and efficient evaporation.
Please note: A vacuum source, vacuum check valve and cooling system are required but not supplied. Please contact your local Avantor sales office for advice and details.
** The valve unit (181-0140) or the Woulff bottle (181-0141) (to be used with a V-700 or V-710 vacuum pump) are available as optional extras.