Labelling tapes for anesthesia drug syringes
Coated paper tape with removable adhesive. The following user applied drug tapes are available for accurate and reliable labelling of all anesthesia drug syringes. This specification covers the size, colour, pattern, and type used on labels applied to unlabelled syringes, filled by the users or their agents, to identify the drug content. In addition, tapes correspond to the published 2004 JCAHO Standard MM,4,30 for appropriately labelling all medications with drug name, strength, amount, expiration date, expiration time, and initials of the person administering the medication.
- Conforms to the nationally recognized ASTM Standard D4774-94(2000) ' Specification for User Applied Drug Labels in Anesthesiology'
: Rolls of 12,7 m - 333 imprints per roll with perforations between.