2-Bromonaphthalene
Catalog # MOLE11563796-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Merck Index:13,01411
Boiling Pt:282 °C (1013 hPa)
Melting Pt:52…55 °C
Density:1.605 g/cm³ (0 °C)
Flash Pt:112 °C
:207.07 g/mol
Storage Temperature:Ambient
EINECS:209-452-5
:GHS07
CAS Number:580-13-2
Formula:C₁₀H₇Br
MDL Number:MFCD00004051