Order Entry
Germany
ContactUsLinkComponent
Zinc oxide ≥99.9% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Environment
Zinc oxide ≥99.9% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 41126.KS
CAS Number:  
Environment
Zinc oxide ≥99.9% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 41126.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :81.39 g/mol
Merck Index:13,10200
Melting Pt:1975 °C
Density:5.61 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-222-5
UN:3077
ADR:9,III
REACH:01-2119463881-32
 :GHS09
CAS Number:1314-13-2
Formula:ZnO
MDL Number:MFCD00011300