Order Entry
Germany
ContactUsLinkComponent
Silver nitrate 99.8-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™
Corrosive
Oxidizing
Environment
Silver nitrate 99.8-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™
Certificates
undefined
Corrosive
Oxidizing
Environment
Silver nitrate 99.8-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™

Specification Test Results

  • Assay:
    99.8 - 100.5 %
  • Acid. or alk. react. impurities:
    complying
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 0.0005 %
  • Lead (Pb):
    Max. 0.002 %
  • Substances not precipitated by HCl (as SO₄):
    Max. 0.03 %
  • Nitrite (NO₂):
    Max. 0.1 %
  • Sulfate (SO₄):
    Max. 0.01 %
  • Al, Pb, Bi and Cu ions:
    complying
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying
  • Vapor Density:
    5.8 (vs air)

Specification Test Results

Related Information

Frequently Bought Together