Lanthanum hexaboride ≥99.5% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12008-21-8
:
Formula:B₆La
MDL Number:MFCD00151350
:203.77 g/mol
Melting Pt:2210 °C
Density:2.61
Storage Temperature:Ambient
EINECS:234-531-6
Related Information
Frequently Bought Together