Xenon difluoride for synthesis, Sigma-Aldrich®
: Merck
:13709-36-9
:
:F₂Xe
:MFCD00040538
:169.29 g/mol
:14,10072
:114 °C (1013 hPa)
:4.32 g/cm³ (20 °C)
:Refrigerator
:237-251-2
:5.1 (8),II
:M-Clarity™ quality level = MQ200
:3085
:GHS03
:GHS06
:GHS05
- Solubility:(20 °C) (decomposition)
- Melting Point:130 - 135 °C (sublimed)
- Molar Mass:169.29 g/mol
- Vapor Pressure:3.8 hPa (25 °C)