


Xenon difluoride ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13709-36-9
:
Formula:F₂Xe
MDL Number:MFCD00040538
:169.29 g/mol
Merck Index:14,10072
Boiling Pt:114 °C (1013 hPa)
Melting Pt:127…129 °C
Density:4.32
Storage Temperature:Ambient
EINECS:237-251-2
UN:3087
ADR:5.1,II
:GHS03
:GHS06
:GHS05