Order Entry
Germany
ContactUsLinkComponent
Xenon difluoride 99.5+%
Corrosive
Oxidizing
Xenon difluoride 99.5+%
Certificates
undefined
Corrosive
Oxidizing
Xenon difluoride 99.5+%

Specification Test Results

  • Appearance:
    White Crystals
  • Purity:
    ≥99.50 %

Specification Test Results

Related Information