
Xenon difluoride
Catalog # ABCRAB104987.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
:169.29 g/mol
Merck Index:14,10072
Boiling Pt:114 °C (1013 hPa)
Density:4.32 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-251-2
UN:3087
ADR:5.1,II
:GHS05
CAS Number:13709-36-9
Formula:F₂Xe
MDL Number:MFCD00040538