
Ammonium nitrate ≥98.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:6484-52-2
:
Formula:NH₄NO₃
MDL Number:MFCD00011425
:80.04 g/mol
Merck Index:13,00538
Boiling Pt:230 °C (1013 hPa)
Melting Pt:169 °C
Density:1.73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:229-347-8
UN:1942
ADR:5.1,III
:GHS03
Specification Test Results
- Assay:Min. 98.0 %
- pH (25°C; 5 %):4.5 - 6.0
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Residue on ignition:Max. 100 ppm
- Cl (Chloride):Max. 3 ppm
- NO2 (Nitrite):Max. 5 ppm
- PO4 (Phosphate):Max. 5 ppm
- SO4 (Sulphate):Max. 20 ppm
- Ca (Calcium):Max. 30 ppm
- Fe (Iron):Max. 2 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500)
Packaging Information
21280_AMMONIUM NITRATE ANALAR NP ACSR.PH.EUR_BSE_REV0.pdf
BSE/TSE Certificates
spec_VWRC21280.260 spec_VWRC21280.260 spec_VWRC21280.293 spec_VWRC21280.293 spec_VWRC21280.460
Product specification
More