Order Entry
Germany
ContactUsLinkComponent
Oxidizing
Irritant
Ammonium nitrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck
Certificates
Oxidizing
Irritant
Ammonium nitrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1920 g/l (20 °C)
  • Melting Point:
    169 °C
  • Molar Mass:
    80.04 g/mol
  • Bulk Density:
    600 - 700 kg/m3
  • Boiling Point:
    302 °C (1013 hPa)
  • Density:
    1.72 g/cm3 (20 °C)
  • pH:
    5.0 - 5.5 (1920 g/l, H2O, 20 °C)
  • Hazard Class:
    5.1
  • Hazard Class Description:
    Oxidizer

Specification Test Results

Related Information

Frequently Bought Together