Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium nitrate
Ammonium nitrate, Supelco®
Ammonium nitrate, Supelco®
:
Merck
Ammonium nitrate, Supelco®
:
Merck
:
6484-52-2
:
:
NH₄NO₃
:
MFCD00011425
:
80.04 g/mol
:
13,00538
:
230 °C (1013 hPa)
:
169 °C
:
1.73 g/cm³ (20 °C)
:
1942
:
5.1,III
:
M-Clarity™ quality level = MQ200
:
229-347-8
:
GHS03
:
GHS07
:
Ambient
MERC1.01187.1000_TDS_EN
ProductPage_101187
msds_101187
msds_101187